Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-XPF, DyLight 405, Clone: 3F2/3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127881V
Catalog No. NBP127881V
Description
Description
XPF Monoclonal antibody specifically detects XPF in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
XPF
XPF 3F2/3
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
ERCC4
A recombinant His-tagged fragment of human XPF protein produced in E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster
Mouse
IgG1
0.2 ml
Cancer, DNA Repair, Nucleotide Excision Repair
Primary
2072