Learn More
anti-ZAP70, Clone: 1E7.2, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP150059
Catalog No. NBP150059
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ZAP70
1E7.2
Unconjugated
PBS (pH 7.4) with 15mM Sodium Azide
70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, SRKFLJ17679, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD
Mouse
IgG1
0.1 mg
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
7535
Human, Mouse
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1:1000, Flow Cytometry 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:50, CyTOF-ready
Purified
ZAP70
A KLH-coupled peptide corresponding to amino acids 282-307 of human ZAP-70
Protein A purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The mouse monoclonal antibody 1E7.2 recognizes ZAP-70, a 70 kDa protein tyrosine kinase expressed in T and NK cells. ZAP-70 is a molecule susceptible to degradation. It is recommended to use freshly prepared cell lysates (protease inhibitors are essential) to avoid non-specific staining of degradation products.