Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOV10L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539378
Description
This gene is similar to a mouse gene that encodes a putative RNA helicase and shows testis-specific expression. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2009].Specifications
MOV10L1 | |
Polyclonal | |
Unconjugated | |
MOV10L1 | |
Csm, Cardiac-specific isoform of Mov10l1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54456, 83456 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BXT6, Q99MV5 | |
MOV10L1 | |
A synthetic peptide derived from the internal region of human MOV10L1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction