Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPG Mouse anti-Human, Clone: 2C5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA519353

Catalog No. PIMA519353


Add to cart

Description

Description

Peptide Sequence: MPARSGAQFC RRMGQKKQRP ARAGQPHSSS DAAQAPAEQP HSSSDAAQAP CPRERCLGPP TTPGPYRSIY FSSPKGHLTR LGLEFFDQPA

3 ft. end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p,CRA36.1 (3-methyl-adenine DNA glycosylase),N-methylpurine-DNA glycosylase, MPG,alkyladenine DNA glycosylase,proliferation-inducing protein 11,proliferation-inducing protein
Specifications

Specifications

MPG
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P29372
MDG; N-methylpurine-DNA glycosylase; AAG; APNG; CRA36.1; Mid1; N-methylpurine-DNA glycosylase, MPG; PIG11; PIG16; anpg; 3′ end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; CRA36.1 (3-methyl-adenine DNA glycosylase); alkyladenine DNA glycosylase; proliferation-inducing protein 11; proliferation-inducing protein 16
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4350
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
2C5
Unconjugated
MPG
Liquid
MPG
MPG (AAH14991, 1 a.a. ∽ 90 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit