Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128215

Catalog No. PA128215

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Myeloperoxidase is a hemoprotein that is abundantly expressed in neutrophils and secreted during their activation. Native Myeloperoxidase is represented as a covalently bound tetrameric complex of two glycosylated alpha chains (MW 59 - 64 kDa) and two unglycosylated beta chains (MW 14 kDa) with total MW 150kDa and theoretical pI 9.2. Traditionally Myeloperoxidase was considered as a main target of anti-neutrophil cytoplasm antibodies (ANCA), the serological markers for certain systemic vasculities e.g. periarteriitis nodosa, microscopic polyarteriitis and pulmonary eosinophilic granulomatosis (Churg-Strauss syndrome). Low to moderate anti-Myeloperoxidase autoantibody levels are also reported in rheumatoid arthritis. Recently it was shown that Myeloperoxidase participates in the initiation and progression of cardiovascular disease. It possesses potent proinflammatory properties and may contribute directly to tissue injury. Now Myeloperoxidase is under consideration as one of the most promising cardiac markers.

PA1-28215 detects MPO from human samples.PA1-28215 has been successfully used in immunohistochemistry (paraffin tissue) applications. Staining of formalin fixed tissues requires boiling tissue sections in 10mM citrate buffer (p.H. 6.0) for 10 minutes followed by cooling at room temperature for 20 minutes.Postive control for PA1-28215 is Tonsil or Spleen.The PA1-28215 immunogen is Purified human granulocytic MPO
Specifications

Specifications

MPO
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P05164
MPO
Purified human granulocytic MPO
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
MPO
Purified
Rabbit
IgG
7mL
Store at 4°C, do not freeze
Primary
4353
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit