Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

membrane protein, palmitoylated 1, 55kDa, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human MPP1 protein.

Manufacturer:  Abnova Corporation H00004354B01

Catalog No. 89-005-782


Add to cart

Description

Description

Palmitoylated membrane protein 1 is the prototype of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. Palmitoylated membrane protein 1 contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. [provided by RefSeq

Sequence: MTLKASEGESGGSMHTALSDLYLEHLLQKRSRPEAVSHPLNTVTEDMYTNGSPAPGSPAQVKGQEVRKVRLIQFEKVTEEPMGITLKLNEKQSCTVARILHGGMIHRQGSLHVGDEILEINGTNVTNHSVDQLQKAMKETKGMISLKVIPNQQSRLPALQMFMRAQFDYDPKKDNLIPCKEAGLKFATGDIIQIINKDDSNWWQGRVEGSSKESAGLIPSPELQEWRVASMAQSAPSEAPSCSPFGKKKKYKDKYLAKHSSIFDQLDVVSYEEVVRLPAFKRKTLVLIGASGVGRSHIKNALLSQNPEKFVYPVPYTTRPPRKSEEDGKEYHFISTEEMTRNISANEFLEFGSYQGNMFGTKFETVHQIHKQNKIAILDIEPQTLKIVRTAELSPFIVFIAPTDQGTQTEALQQLQKDSEAIRSQYAHYFDLSLVNNGVDETLKKLQEAFDQACSSPQWVPVSWVY
Specifications

Specifications

membrane protein, palmitoylated 1, 55kDa
Unconjugated
No additive
NM_002436.2
AAG12|DXS552|DXS552E|EMP55|MRG1|PEMP
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4354
Western Blot
Mouse polyclonal antibody raised against a full-length human MPP1 protein.
MPP1
Serum
MPP1
MPP1 (NP_002427.1, 1 a.a. ~ 466 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit