Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MPP2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132393

Catalog No. NBP132393


Add to cart

Description

Description

MPP2 Polyclonal antibody specifically detects MPP2 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

MPP2
0.61mg/mL
Western Blot 1:500-1:3000
Affinity Purified
MPP2
Recombinant protein encompassing a sequence within the center region of human MPP2. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
4355
Human, Mouse
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Discs large homolog 2, DKFZp686A06252, DKFZp686J2189, DKFZp761D0712, DLG2discs large, homolog 2, MAGUK p55 subfamily member 2, membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2), Protein MPP2
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only