Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

metallophosphoesterase domain containing 1 (B02), Mouse anti-Human, Rat, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human MPPED1 protein.

Manufacturer:  Abnova Corporation H00000758B02

Catalog No. 89-005-012


Add to cart

Description

Description

Sequence: MWRSRWDASVLKAEALALLPCGLGMAFSQSHVMAARRHQHSRLIIEVDEYSSNPTQAFTFYNINQGRFQPPHVQMVDPVPHDAPKPPGYTRFVCVSDTHSRTDPIQMPYGDVLIHAGDFTELGLPSEVKKFNEWLGSLPYEYKIVIAGNHELTFDQEFMADLIKQDFYYFPSVSKLKPENYENVQSLLTNCIYLQDSEVTVRGFRIYGSPWQPWFYGWGFNLPRGQALLEKWNLIPEGVDILITHGPPLGFLDWVPKKMQRVGCVELLNTVQRRVQPRLHVFGHIHEGYGVMADGTTTYVNASVCTVNYQPVNPPIVIDLPTPRNS
Specifications

Specifications

metallophosphoesterase domain containing 1
Unconjugated
No additive
NM_001585.2
239AB|C22orf1|FAM1A|MGC88045
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
758
Immunohistochemistry (PFA fixed), Western Blot
Mouse polyclonal antibody raised against a full-length human MPPED1 protein.
MPPED1
Serum
MPPED1
MPPED1 (NP_001576.3, 1 a.a. ~ 326 a.a) full-length human protein.
RUO
Y
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit