Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M6PR Chicken anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129512

Catalog No. PA129512


Add to cart

Description

Description

PA1-29512 detects MPR46 from mouse, human, rat samples. PA1-29512 has been successfully used in immunocytochemistry and Western blot applications. The PA1-29512 immunogen is: Synthetic peptide corresponding to amino acids 60-114 of Human Mannose 6 Phospate Receptor. Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.

The protein encoded by this gene is a receptor for mannose-6-phosphate groups on lysosomal enzymes. The receptor forms a homodimer or homotetramer for intracellular targeting of lysosomal enzymes and export of newly synthesized lysosomal enzymes into the cell secretions. The receptor is an integral membrane protein which localizes to the trans-Golgi reticulum, endosomes, and the plasma membrane.
Specifications

Specifications

M6PR
Polyclonal
Unconjugated
M6PR
Liquid
M6PR
synthetic peptide corresponding to amino acids 60-114 of Human Mannose 6 Phospate Receptor.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
2 mg/mL
PBS with 0.02% sodium azide
P20645, P24668, Q6AY20
CD-MPR, FLJ32994, MPR46, SMPR
Chicken
IgY
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17113, 312689, 4074
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit