Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mps1 Mouse anti-Human, Clone: 4-112-3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05683
Description
Anti-Mps1 Antibody, CT, clone 4-112-3 is a high quality Mouse Monoclonal Antibody for the detection of Mps1 and has been validated in immunoprecipitation, western blot, immunocytochemistry.Specifications
Mps1 | |
Monoclonal | |
Unconjugated | |
NM_003318 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C for 24 months. | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
4-112-3 | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
TTK, PYT, MPS1L1, ESK, FLJ38280 | |
His-tagged fusion protein corresponding to amino acids 3-856 of human Mps1 | |
200 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction