Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPS1, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131888

Catalog No. PA131888

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes an enzyme that hydrolyzes the terminal alpha-L-iduronic acid residues of two glycosaminoglycans, dermatan sulfate and heparan sulfate. This hydrolysis is required for the lysosomal degradation of these glycosaminoglycans. Mutations in this gene that result in enzymatic deficiency lead to the autosomal recessive disease mucopolysaccharidosis type I (MPS I).

PA1-31888 detects MPS1 from human samples.PA1-31888 has been successfully used in Western blot applications.The PA1-31888 immunogen is: Synthetic peptide: PLAKDLLHPSPEE from N Terminus of the protein sequence according to NP_001021.
Specifications

Specifications

MPS1
Polyclonal
Unconjugated
RPS27
Purified
RPS27
synthetic peptide PLAKDLLHPSPEE from N Terminus of the protein sequence according to NP_001021.
IgG fraction
RUO
Antibody
Polyclonal
Human, Rat
In Situ Hybridization (ISH), Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide
P42677, Q71TY3
IDA, MPS1
Goat
IgG
100μg
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
6232
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit