Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MPV17, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152097

Catalog No. NBP152097


Add to cart

Description

Description

MPV17 Polyclonal antibody specifically detects MPV17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.
Specifications

Specifications

MPV17
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Peptide ELISA Detection limit 1:32000
NP_002428.1
human homolog of glomerulosclerosis and nephrotic syndrome, MpV17 mitochondrial inner membrane protein, murine homolog, glomerulosclerosis, protein Mpv17
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
ELISA, Immunohistochemistry, Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
MPV17
Peptide with sequence C-QRALAAHPWKVQ corresponding to internal region according to NP_002428.1.
Immunogen affinity purified
RUO
Primary
4358
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only