Learn More
MRAP Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526979
Catalog No. PIPA526979
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MRAP encodes a melanocortin receptor-interacting protein. The encoded protein regulates trafficking and function of the melanocortin 2 receptor in the adrenal gland. The encoded protein can also modulate signaling of other melanocortin receptors. Mutations in this gene have been associated with familial glucocorticoid deficiency type 2.
Specifications
Specifications
MRAP
Polyclonal
Unconjugated
MRAP
Liquid
MRAP
KLH conjugated synthetic peptide between 7-36 amino acids from the N-terminal region of human MRAP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8TCY5
B27, C21orf61, FALP, FGD2, GCCD2, fat cell-specific low molecular weight protein, fat tissue-specific low MW protein, melanocortin-2 receptor accessory protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56246