Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mre11 Mouse, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611366

Catalog No. BDB611366


Add to cart

Description

Description

Originally identified in S. cerevisiae, Rad50 is one of a group of genes, designated the Rad52 epistasis group, whose products mediate DNA double-stranded break (DSB) repair. Many of these genes, including Rad50, are conserved in humans and have a similar function to their S. cerevisiae counterparts. In yeast, a multiprotein complex of Rad50, Mre11, and XRS2 has been implicated in the nucleocytic processing of DSBs, in homologous recombination, in nonhomologous end joining, and in telomere maintenance. In humans, Rad50 and Mre11 complex with up to three additional proteins (95kDa, 200kDa, and 350kDa). The 95kDa species is thought to be human XRS2, although a separate report has identified it as Nibrin, the product of the gene mutated in Nijmegen breakage syndrome. The Rad50-Mre11-p95 complex possesses endonuclease and 3 ft. to 5 ft. exonuclease activity. Mre11 may provide both the endonuclease and exonuclease activity, but may also be involved in other functions related to this complex. Mre11 is ubiquitously expressed, with the highest expression in proliferating tissues. Thus, MRE11 may function in a multiprotein complex that has many roles during DNA maintenance and repair.

Immunofluorescence, Western Blotting

Specifications

Specifications

Mre11
18
Unconjugated
Affinity Purified
Human Mre11 aa. 3-194
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit