Learn More
MRGPRX3 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00111W
Catalog No. PIOSR00111W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Orphan receptor activated by a subset of RFamide-family neuropeptides such as FLRF-amide and FMRF-amide. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system. Its effect is mediated by G(q) and G(11) proteins. May regulate the function of nociceptive neurons by modulation of pain perception. Expressed in a subset of IB4-positive small diameter nociceptive dorsal root neurons.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MRGPRX3
Polyclonal
Mrgprx3
Nuclear receptor MrgA10 RF-amide G protein-coupled receptor, Mrga10, MrgA, Mrgpra, MGC108580
Rabbit
100 μL
Primary
Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q7TN49
MRGPRX3
A synthetic peptide from the 4th cytoplasmic loop of rat Mas-related G-protein coupled receptor member A
RUO
252960
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations