Learn More
MRGD Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00106W
Catalog No. PIOSR00106W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability.
May regulate nociceptor function and/or development, including the sensation or modulation of pain. Functions as a specific membrane receptor for beta-alanine. Beta-alanine at micromolar doses specifically evoked Ca(2+) influx in cells expressing the receptor. Beta-alanine decreases forskolin-stimulated cAMP production in cells expressing the receptor, suggesting that the receptor couples with G-protein G(q) and G(i). Localized at the plasma membrane but internalized into the cytoplasm after treatment with beta-alanine.
Specifications
Specifications
MRGD
Polyclonal
whole serum with no preservative
Q8TDS7
Mas-related G-protein coupled receptor, member D; Beta-alanine receptor, G-protein coupled receptor TGR7, MRGD
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
MRGPRD
Lyophilized
MRGPRD
A synthetic peptide from the 4th cytoplasmic loop of human Mas-related G-protein coupled receptor member D
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
116512