Learn More
MRGPRE Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00107W
Catalog No. PIOSR00107W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Orphan receptor. May regulate nociceptor function and/or development, including the sensation or modulation of pain.
Specifications
Specifications
MRGPRE
Polyclonal
whole serum with no preservative
Q86SM8
Mas-related G-protein coupled receptor, member E; G-protein coupled receptor 167, GPR167, MRGE, MGC138408
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Non-human Primate
Immunohistochemistry, Western Blot
Unconjugated
MRGPRE
Lyophilized
MRGPRE
A synthetic peptide from the 3d cytoplasmic loop of human Mas-related G-protein coupled receptor member E
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
116534