Learn More
MRGPRX2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00110W
Catalog No. PIOSR00110W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Orphan receptor. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Cortistatin-14 seems to be a high potency ligand at this receptor. Cortistatin has several biological functions including roles in sleep regulation locomotor activity, and cortical function. In receptor-expressing cells, cortistatin-stimulated increases in intracellular Ca(2+) but had no effect on basal or forskolin-stimulated cAMP levels, suggesting that this receptor is G(q)-coupled. Has a limited expression profile, both peripheral and within the central nervous system, with highest levels in dorsal root ganglion.
Specifications
Specifications
MRGPRX2
Polyclonal
whole serum with no preservative
Q96LB1
Mas-related G-protein coupled receptor member X2, MRGPRX2, MAS-related GPR, member X2
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
MRGPRX2
Lyophilized
MRGPRX2
A synthetic peptide from the 4th cytoplasmic loop of human Mas-related G-protein coupled receptor member X2
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
117194