Learn More
anti-MRP1, Clone: MRPm5, Invitrogen™
Supplier: Thermo Scientific MA135675
Description
Host Species: Mouse
Clone: MRPm5
Cross Reactivity: Hu
Immunogen: Bacterial fusion protein of MRP1
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=
Fluorescence Activated Cell Sorting, Immunocytochemistry, Immunohistochemistry (Frozen/Paraffin), Western Blotting

Specifications
MRP1 | |
MRPm5 | |
Mouse | |
1 mL | |
Human |
Monoclonal | |
Unconjugated | |
Bacterial fusion protein of MRP1 | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.