Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A9 Mouse anti-Human, Clone: 1H9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA140222

Catalog No. 01-676-562


Add to cart

Description

Description

MA1-40222 detects MRP-14 from human samples. MA1-40222 has been successfully used in ELISA, flow cytometry and Western blot applications. The MA1-40222 immunogen is KPpL epitope on human MRP-14.

MRP-14 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
Specifications

Specifications

S100A9
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P06702
S100A9
Mouse
IgG1
100 μg
4° C
Primary
6280
ELISA, Flow Cytometry, Western Blot
1H9
Unconjugated
S100A9
Liquid
S100A9
KPpL epitope on Human MRP-14.
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit