Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MRPL36, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192139

Catalog No. NBP192139


Add to cart

Description

Description

MRPL36 Polyclonal antibody specifically detects MRPL36 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

MRPL36
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
BRCA1-interacting protein 1, BRIP1MGC104245, L36mtMRP-L36PRPL36, mitochondrial ribosomal protein L36,39S ribosomal protein L36, mitochondrial, putative BRCA1-interacting protein, RPMJ
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Affinity Purified
MRPL36
This antibody was developed against Recombinant Protein corresponding to amino acids:PVAVEPGAAVRSLLSPGLLPHLLPALGFKNKTVLKKRCKDCYLVKRRGRWYVYCKTHPRHKQRQM
Immunogen affinity purified
RUO
Primary
64979
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only