Learn More
anti-MRPS18B, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP215024
Catalog No. NBP215024
Add to cart
Description
Description
MRPS18B Polyclonal specifically detects MRPS18B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
MRPS18B
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
MRPS18B
Recombinant protein encompassing a sequence within the center region of human MRPS18B. The exact sequence is proprietary.
0.1 mL
Core ESC Like Genes, Stem Cell Markers
28973
Human
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.84mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
28S ribosomal protein S18-2, mitochondrial, C6orf1428S ribosomal protein S18b, mitochondrial, HSPC183, HumanS18a, mitochondrial ribosomal protein S18-2, mitochondrial ribosomal protein S18B, MRP-S18-2, MRPS18-2DKFZp564H0223, Mrps18-b, MRP-S18-b, PTD017, S18amt, S18mt-b
Rabbit
Affinity Purified
RUO
Primary
Human. Predicted cross-reactivity based on sequence identity: Monkey (94%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.