Learn More
MRPS24 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523999
Catalog No. PIPA523999
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPS24 encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 11.
Specifications
Specifications
MRPS24
Polyclonal
Unconjugated
MRPS24
Liquid
MRPS24
KLH conjugated synthetic peptide between 21-50 amino acids from the central region of human MRPS24
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.35 mg/mL
PBS with 0.09% sodium azide
Q96EL2
MRP-S24, S24mt, bMRP-47, bMRP47, 28S ribosomal protein S24, mitochondrial, mitochondrial 28S ribosomal protein S24
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64951