Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MS A+G+M (H+L) AB BIOTIN  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52600049
Affinity purified polyclonal antibody to Mouse IgA, IgG, and IgM, heavy and light chains (whole IgA, IgG, IgM) made in Goat and biotinylated. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-77