Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MS G+M (H+L) AB DL488  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300392
Affinity purified polyclonal antibody to Mouse IgG and IgM, both heavy and light chains (whole IgG and IgM) made in Goat and labeled with DyLight 488. Cross-reactivity with human serum has been minimized with affinity procedures. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-86