Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MS IGA (A) AB HSA HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200367
Affinity purified polyclonal antibody to Mouse IgA heavy chains (alpha) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-29