Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MS IGG (G) AB HSA HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200460
Affinity purified polyclonal antibody to Mouse IgG heavy chains (gamma) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-90