Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MS IGG(H+L)AB RT+HSA FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300310
Affinity purified polyclonal secondary antibody against Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross-adsorbed with human serum.
Catalog No. 50-668-96