Learn More
Seracare Life Sciences Inc ANTI-MS IGG(H+L) FAB 2 BIOTIN
Supplier:  Seracare Life Sciences Inc 52600064
Encompass
Affinity purified polyclonal antibody to the F(ab)2 fragment of Mouse IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-75
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.