Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MS IGG (H+L) HSA AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200286
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin.
Catalog No. 50-669-41