Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MS IGM (U) AB HSA BIOTIN  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52600047
Affinity purified polyclonal antibody to Mouse IgM heavy chains (mu) made in Goat and biotinylated. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-75