Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A4B Rat anti-Mouse, Clone: 444008, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524168

Catalog No. PIMA524168


Add to cart

Description

Description

This antibody stains mouse MS4A4B transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

MS4A4B
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Lyophilized
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
60361
Flow Cytometry
444008
Unconjugated
Ms4a4b
Ms4a4b
HEK293 human embryonic kidney cell line transfected with mouse MS4A4B Met1-Val226
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit