Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10056428
Description
MSH2 Polyclonal specifically detects MSH2 in Human, Mouse samples. It is validated for Western Blot.Specifications
MSH2 | |
Polyclonal | |
Unconjugated | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
MSH2 | |
A portion of amino acids 550-600 of human MSH2 was used as the immunogen. | |
0.1 mg | |
Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors | |
4436 | |
Human, Mouse | |
Purified |
Western Blot | |
0.5 mg/mL | |
Western Blot 1-3 ug/ml | |
COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human, dog and S. cerevisiae, 94% in mouse and rat, and 76% in zebrafish. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction