Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527449

Catalog No. PIPA527449


Add to cart

Description

Description

PA5-27449 targets MSI1 in IF and WB applications and shows reactivity with Human samples. The PA5-27449 immunogen is recombit fragment corresponding to a region within amino acids 176 and 362 of Human MSI1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a protein containing two conserved tandem RNA recognition motifs. Similar proteins in other species function as RNA-binding proteins and play central roles in posttranscriptional gene regulation. Expression of this gene has been correlated with the grade of the maligcy and proliferative activity in gliomas and melanomas. A pseudogene for this gene is located on chromosome 11q13.
Specifications

Specifications

MSI1
Polyclonal
Unconjugated
MSI1
Liquid
MSI1
Recombit fragment corresponding to a region within amino acids 176 and 362 of Human MSI1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
0.69 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O43347, Q61474, Q8K3P4
RNA-binding protein Musashi homolog 1, musashi-1, musashi1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17690, 259272, 4440
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit