Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-MSIGG(H+L)F(AB )2 HSA HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200395
Affinity purified polyclonal antibody, F(ab)2 fragment, to Mouse IgG, heavy and light chains (whole IgG).;Made in Goat and labeled with horseradish peroxidase. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-66