Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MSK2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP146187

Catalog No. NBP146187


Add to cart

Description

Description

MSK2 Polyclonal antibody specifically detects MSK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

MSK2
0.2 mg/mL
Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2 - 5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500
Affinity Purified
RPS6KA4
The immunogen for this product maps to a region between residue 722 and 772 of human Mitogen- and Stress-activated Protein Kinase 2 using the numbering given in entry NP_003933.1 (GeneID 8986).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
ribosomal protein S6 kinase, 90kDa, polypeptide 4
Rabbit
IgG
0.1 ml
Protein Kinase
Primary
8986
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only