Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MST1/2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128672

Catalog No. NBP128672


Add to cart

Description

Description

MST1/2 Polyclonal antibody specifically detects MST1/2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MST1/2
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
STK3
The immunogen this antibody was made to, maps to the C-terminus of human Mammalian Sterile 20-like 2 (Serine/Threonine Kinase 3) using the numbering given in Swiss Prot entry Q13188 (GeneID 6788). The epitope is highly conserved with MST1(STK4).
Immunogen affinity purified
RUO
Primary
6788
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500
Q13188
D3F15S2, DNF15S2, HGFL, MSP, MST1 macrophage stimulating 1 (hepatocyte growth factor-like), NF15S2
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only