Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MST1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141410

Catalog No. 01-675-539


Add to cart

Description

Description

This antibody will cross-react with mouse and rat MST-1, based on sequence homology. Suggested positive control: Jurkat whole cell lysate.

Mammalian STE20-like kinase 1, or MST-1, is a serine/threonine kinase that, upon cleavage, has been implicated in the promotion of chromatin condensation. MST-1 is also known as KRS2 and STK4. The C-terminus of MST-1 contains two functional NESs (nuclear export signals), which are released upon caspase-mediated cleavage. The N-terminus portion of the protein then translocates to the nucleus and will promote chromatin condensation at sufficiently high levels. Full-length MST-1 is localized to the cytoplasm.
Specifications

Specifications

MST1
Polyclonal
Unconjugated
STK4
Liquid
STK4
Synthetic peptide corresponding to residues A(372) E D E E E E G T M K R R D E T M Q P(390) of human MST-1.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q13043
KRS2,STK4
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6789
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit