Learn More
anti-MST2/STK3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232765
Catalog No. NBP232765
Add to cart
Description
Description
MST2/STK3 Polyclonal antibody specifically detects MST2/STK3 in Human, Zebrafish samples. It is validated for Western Blot.
Specifications
Specifications
MST2/STK3
0.5 mg/mL
Western Blot 1:1000
Q7ZUQ3
EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2
Rabbit
IgG
Protein A purified
RUO
Primary
6788
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
STK3
This DANRE stk3(36kDa subunit) antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 228-252 amino acids from the Central region of DANRE stk3(36kDa subunit).
56.081 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Zebrafish