Learn More
MST3 Mouse, Unlabeled, Clone: 47, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 611056
Description
Ste20 is a S. cerevisiae Ser/Thr protein kinase that functions upstream of the MAP kinase module. Mammalian and yeast homologs of this kinase are divided into two classes based on their structure and regulation. Members of the first class (Ste20, Cla4, and p21-activated protein kinase [PAK]) contain a C-terminal kinase domain, an N-terminal regulatory domain and a small GTPase Rac1/Cdc42 binding domain. Members of the second class lack GTPase binding sites, but are similar to the former class throughout the catalytic domain. The latter class includes GC kinase, HPK, KHS, KRS1 & 2, MST1, 2, & 3, and SOK-1. MST3 (Mammalian Sterile Twenty-like) is a ubiquitous cytoplasmic kinase that contains an N terminal kinase domain and a C-terminal regulatory domain. It shares extensive sequence similarity with SOK-1 and is similarly autophosphorylated and activated. MST3 remains distinct in that it prefers Mn2+ to Mg2+ as a divalent cation and uses GTP and ATP as phosphate donors. It does not respond to a variety of mitogenic and stress stimuli and fails to activate known mammalian MAPK pathways. Although its function is undefined, MST3 may function in a novel intracellular signaling pathway.
Immunofluorescence, Western Blotting
Specifications
MST3 | |
Monoclonal | |
250μg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgG1 |
Immunofluorescence, Western Blot | |
47 | |
Unconjugated | |
Human MST3 aa. 275-393 | |
50 μg | |
Cell Biology | |
Human, Mouse, Rat |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.