Learn More
MST3 Mouse, Unlabeled, Clone: 47, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 611057
 View more versions of this product
Catalog No. BDB611057
Add to cart
Description
Description
Ste20 is a S. cerevisiae Ser/Thr protein kinase that functions upstream of the MAP kinase module. Mammalian and yeast homologs of this kinase are divided into two classes based on their structure and regulation. Members of the first class (Ste20, Cla4, and p21-activated protein kinase [PAK]) contain a C-terminal kinase domain, an N-terminal regulatory domain and a small GTPase Rac1/Cdc42 binding domain. Members of the second class lack GTPase binding sites, but are similar to the former class throughout the catalytic domain. The latter class includes GC kinase, HPK, KHS, KRS1 & 2, MST1, 2, & 3, and SOK-1. MST3 (Mammalian Sterile Twenty-like) is a ubiquitous cytoplasmic kinase that contains an N terminal kinase domain and a C-terminal regulatory domain. It shares extensive sequence similarity with SOK-1 and is similarly autophosphorylated and activated. MST3 remains distinct in that it prefers Mn2+ to Mg2+ as a divalent cation and uses GTP and ATP as phosphate donors. It does not respond to a variety of mitogenic and stress stimuli and fails to activate known mammalian MAPK pathways. Although its function is undefined, MST3 may function in a novel intracellular signaling pathway.
Immunofluorescence, Western Blotting
Specifications
Specifications
MST3
47
Unconjugated
Mouse
IgG1
RUO
Primary
Human, Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Human MST3 aa. 275-393
150μg
Cell Biology
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.