Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
MSX1 Mouse anti-Human, Clone: 1D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519366
Catalog No. PIMA519366
Description
Description
Peptide Sequence: NRRAKAKRLQ EAELEKLKMA AKPMLPPAAF GLSFPLGGPA AVAAAAGASL YGASGPFQRA ALPVAPVGLY TAHVGYSMYH LT
This gene encodes a member of the muscle segment homeobox gene family. The encoded protein functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. It may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in this gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal domit hypodontia.
Specifications
Specifications
MSX1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
P28360
HOX7, HYD1, STHAG1, homeobox 7, homeobox protein Hox-7, homeobox protein MSX-1, msh homeo box 1, msh homeobox 1-like protein, msh homeobox homolog 1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4487
ELISA, Immunoprecipitation, Western Blot
1D2
Unconjugated
MSX1
Liquid
MSX1
MSX1 (NP_002439, 216 a.a. ∽ 297 a.a) full length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human