Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | MSX1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 2.5-5 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224715SS
|
Novus Biologicals
NBP224715SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP224715
|
Novus Biologicals
NBP224715 |
0.1 mg |
Each for $482.50
|
|
|||||
Description
MSX1 Polyclonal specifically detects MSX1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MSX1 | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 2.5-5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
Homeobox protein Hox-7, homeobox protein MSX-1, HOX7homeobox 7, HYD1msh homeobox homolog 1 (Drosophila), msh (Drosophila) homeo box homolog 1 (formerly homeo box 7), msh homeo box 1, msh homeobox 1, Msh homeobox 1-like protein, msh homeobox homolog 1, OFC5, STHAG1 | |
MSX1 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, mouse and rat, 83% homologous in opossum and monkey and 73% homologous in dog. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Primate | |
P28360 | |
4487 | |
A portion of amino acids 50-100 of human HOX7 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title