Learn More
MTA3 Mouse anti-Human, Clone: 3B3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522371
Catalog No. PIMA522371
Add to cart
Description
Description
Peptide Sequence: LKMPTQSEEE KLSPSPTTED PRVRSHVSRQ AMQGMPVRNT GSPKSAVKTR QAFFLHTTYF TKFARQVCKN TLRLRQAARR PFVAINYAAI RAECKMLLNS
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.
Specifications
Specifications
MTA3
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9BTC8
metastasis associated family, member 3, metastasis associated gene family, member 3, metastasis-associated protein MTA3
Mouse
IgG2b, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
57504
ELISA, Immunocytochemistry, Immunofluorescence
3B3
Unconjugated
MTA3
Liquid
MTA3
MTA3 (NP_065795, 416 a.a. ∽ 515 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human