Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTCP1 Monoclonal Antibody (1G12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA519372
Description
Peptide Sequence: MPQKDPCQKQ ACEIQKCLQA NSYMESKCQA VIQELRKCCA QYPKGRSVVC SGFEKEEEEN LTRKSASK
MTCP1 Monoclonal antibody specifically detects MTCP1 in Human samples. It is validated for ELISA, Western Blot.Specifications
MTCP1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P56278 | |
MTCP1 | |
MTCP1 (AAH02600, 1 a.a. ∽ 68 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
1G12 | |
Unconjugated | |
MTCP1 | |
C6.1b; C61B; Cmc4; mature T cell proliferation 1; mature T-cell proliferation 1; mature T-cell proliferation 1 isoform p13; mature T-cell proliferation 1 neighbor protein; mature T-cell proliferation-1 type A; mature T-cell proliferation-1 type B1; MTCP1; MTCP-1 type A; MTCP-1 type B1; p13MTCP1; p8MTCP1; Protein p13 MTCP-1; protein p8 MTCP-1 | |
Mouse | |
Affinity chromatography | |
RUO | |
4515 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction