Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTGR1, Polyclonal, Invitrogen™

Catalog No. PIPA130657
Change view
Click to view available options
Quantity:
250 μg
Catalog No. Quantity
PIPA130657 250 μg
1 options

Catalog No. PIPA130657

Supplier: Thermo Scientific PA130657

Sheep Polyclonal Antibody

Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as

PA1-30657 detects MTGR1 from human samples.PA1-30657 has been successfully used in Western blot applications.The PA1-30657 immunogen is: Unfortunately, this information is considered to be commercially sensitive
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTGR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene CBFA2T2
Gene Accession No. O43439
Gene Alias DKFZp313F2116, EHT, ZMYND3 CBFA2T2
Gene Symbols CBFA2T2
Host Species Sheep
Immunogen Unfortunately, this information is considered to be commercially sensitive
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.