Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTMR15 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525171

Catalog No. PIPA525171


Add to cart

Description

Description

MTMRF encodes a member of the myotubularin-related class 1 cysteine-based protein tyrosine phosphatases. The encoded protein may be catalytically inactive.
Specifications

Specifications

MTMR15
Polyclonal
Unconjugated
FAN1
Liquid
FAN1
KLH conjugated synthetic peptide between 146-174 amino acids from the N-terminal region of human MTMR15
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9Y2M0
KIAA1018, MTMRF, coiled-coil domain-containing protein MTMR15, fanconi anemia associated nuclease 1, fanconi-associated nuclease 1, myotubularin related protein 15, myotubularin-related protein 15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22909
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit