Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mTOR Mouse anti-Human, Clone: 21A12.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051564
Description
Detect mTOR using this Anti-mTOR Antibody, clone 21A12.2 validated for use in western blotting and immunocytochemistry.Specifications
mTOR | |
Monoclonal | |
Unconjugated | |
NP_004949.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
21A12.2 | |
Purified mouse monoclonal in buffer containing 0.1MTris-Glycine (pH7.4), 150mM NaCl, with 0.05% sodium azide. | |
DKFZp686B11164, KIAA1999, MGC39830, mAVO3, pianissimo | |
Full-length human mTOR | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only