Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mTOR Mouse anti-Human, Mouse, Clone: 2ID8.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051592
Description
Anti-mTOR Antibody, clone 2ID8.2 is an antibody against mTOR for use in western blotting and immunocytochemistry.Specifications
mTOR | |
Monoclonal | |
Unconjugated | |
NP_004949.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for up to 12 months. | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
2ID8.2 | |
Purified ascites containing 0.1MTris-Glycine, pH 7.4, 150mM NaCl and 0.05% Sodium Azide. | |
FLJ44809, FRAP, FRAP1, FRAP2, OTTHUMP00000001983, RAFT1, RAPT1, mTOR | |
Flag-tagged recombinant protein corresponding to residues surrounding mTOR. | |
100 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only