Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-mTOR Oligoclonal, Invitrogen™

mTOROligoclonal antibody 710408 from Thermo Scientific™ Pierce™ specifically detects mTOR in Human and Rat samples, and it is validated for WB, IF, ICC, FACS and ChIP

Manufacturer:  Invitrogen 710408

Encompass

Catalog No. 71-040-8


Add to cart

Description

Description

Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation.

710408 detects mTOR in Human and Rat samples. 710408 has been successfully used in WB, IF, ICC, FACS and ChIP procedures. The immunogen for 710408 is recombinant protein corresponding to amino acids 1126-1411 of human mTOR.

Applications:

Chromatin Immunoprecipitation, FACS, Immunocytochemistry, Immunofluorescence, Western Blotting

Specifications

Specifications

mTOR
Unconjugated
P42345, P42346
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Polyclonal
Human, Rat
7HCLC
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
Recombinant protein corresponding to amino acids 1126-1411 of human mTOR.
Protein A
Primary
2475, 56718
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit